Order Entry
Italy
ContactUsLinkComponent
4-[4-(Hydroxymethyl)pyridin-2-yl]morpholine 97%
4-[4-(Hydroxymethyl)pyridin-2-yl]morpholine 97%
Catalog # APOSOR9782-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-[4-(Hydroxymethyl)pyridin-2-yl]morpholine 97%
Catalog # APOSOR9782-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :194.23 g/mol
Storage Temperature:Ambient
CAS Number:556109-99-0
Formula:C₁₀H₁₄N₂O₂
MDL Number:MFCD08271915