

2-Bromo-(5-bromo-1-benzofuran-2-yl)ethan-1-one 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7039-76-1
:
Formula:C₁₀H₆Br₂O₂
MDL Number:MFCD06658982
:317.961 g/mol
Melting Pt:135…136 °C
UN:3261
ADR:8,II
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together