Order Entry
Italy
ContactUsLinkComponent
 
2-Amino-3-methyl-3H-imidazole-4-carboxylic acid ethyl ester 98%
Catalog # APOSOR963687-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Amino-3-methyl-3H-imidazole-4-carboxylic acid ethyl ester 98%
Catalog # APOSOR963687-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Ethyl 2-amino-1-methyl-1H-imidazole-5-carboxylate
 :
 :169.181 g/mol
Storage Temperature:Ambient
CAS Number:177760-04-2
Formula:C₇H₁₁N₃O₂
MDL Number:MFCD09997799