Order Entry
Italy
ContactUsLinkComponent
2-(Chloromethyl)morpholine hydrochloride 97%
2-(Chloromethyl)morpholine hydrochloride 97%
Catalog # APOSOR963189-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Chloromethyl)morpholine hydrochloride 97%
Catalog # APOSOR963189-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :172.053 g/mol
Storage Temperature:Ambient
CAS Number:144053-97-4
Formula:C₅H₁₀ClNO·ClH
MDL Number:MFCD18711391