2-(tert-Butoxycarbonyl)-5,7-dichloro-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:851784-82-2
:
Formula:C₁₅H₁₇Cl₂NO₄
MDL Number:MFCD22370640
:346.206 g/mol
Storage Temperature:Ambient
Frequently Bought Together