
1-tert-Butyl 2-methyl 5-oxopiperidine-1,2-dicarboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:869564-40-9
:
Formula:C₁₂H₁₉NO₅
MDL Number:MFCD15071863
:257.283 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together