Order Entry
Italy
ContactUsLinkComponent
2,6-Diazaspiro[3.4]octan-5-one hydrochloride
2,6-Diazaspiro[3.4]octan-5-one hydrochloride
Catalog # APOSOR960531-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Diazaspiro[3.4]octan-5-one hydrochloride
Catalog # APOSOR960531-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :162.617 g/mol
Storage Temperature:Ambient
CAS Number:1203683-28-6
Formula:C₆H₁₀N₂O·ClH
MDL Number:MFCD11053741