
4-(Aminomethyl)phenol, N-BOC protected 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:149505-94-2
:
Formula:C₁₂H₁₇NO₃
MDL Number:MFCD18206146
:223.268 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together