Vai a Intestazione
Vai a Contenuti principale
Vai a Piè di pagina
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dichloro-2-methylphenol
4,5-Dichloro-2-methylphenol 98%
4,5-Dichloro-2-methylphenol 98%
4,5-Dichloro-2-methylphenol 98%
APOSOR957601-1G
:
Apollo Scientific
:
4,5-Dichloro-2-methylphenol 98%
APOSOR957601-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
177.028 g/mol
:
Ambient
:
52780-67-3
:
C₇H₆Cl₂O
:
MFCD18970307