Order Entry
Italy
ContactUsLinkComponent
3-(2-Methoxyethoxy)-4-methylphenol 95%
3-(2-Methoxyethoxy)-4-methylphenol 95%
Catalog # APOSOR957580-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(2-Methoxyethoxy)-4-methylphenol 95%
Catalog # APOSOR957580-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :182.216 g/mol
Storage Temperature:Ambient
CAS Number:1395035-72-9
Formula:C₁₀H₁₄O₃
MDL Number:MFCD23110693