Order Entry
Italy
ContactUsLinkComponent
 
3-(Propan-2-ylsulphanyl)phenol 95%
Catalog # APOSOR957499-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Propan-2-ylsulphanyl)phenol 95%
Catalog # APOSOR957499-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.256 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₂OS
MDL Number:MFCD24721312