Order Entry
Italy
ContactUsLinkComponent
3-Methoxy-4-methylphenol 95%
3-Methoxy-4-methylphenol 95%
Catalog # APOSOR957439-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Methoxy-4-methylphenol 95%
Catalog # APOSOR957439-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :138.164 g/mol
Storage Temperature:Ambient
CAS Number:19217-50-6
Formula:C₈H₁₀O₂
MDL Number:MFCD16998731