Order Entry
Italy
ContactUsLinkComponent
Propan-2-yl 3-chlorobenzoate 95%
Propan-2-yl 3-chlorobenzoate 95%
Catalog # APOSOR957153-1G
Supplier:  Apollo Scientific
CAS Number:  
Propan-2-yl 3-chlorobenzoate 95%
Catalog # APOSOR957153-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198.646 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₁ClO₂
MDL Number:MFCD04329630