Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Chloro-2-(cyclopropylmethoxy)benzene
1-Chloro-2-(cyclopropylmethoxy)benzene 95%
1-Chloro-2-(cyclopropylmethoxy)benzene 95%
1-Chloro-2-(cyclopropylmethoxy)benzene 95%
APOSOR957057-1G
:
Apollo Scientific
:
1-Chloro-2-(cyclopropylmethoxy)benzene 95%
APOSOR957057-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
182.647 g/mol
:
Ambient
:
1281653-80-2
:
C₁₀H₁₁ClO
:
MFCD17292492