(R)-Methyl 2-methoxy-2-phenylacetate 95%
Catalog # APOSOR955653-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.2 g/mol
Storage Temperature:Ambient
CAS Number:32174-46-2
Formula:C₁₀H₁₂O₃
MDL Number:MFCD16879060