Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(2-Pyridinyl)-2-propenoic acid methyl ester
3-(2-Pyridinyl)-2-propenoic acid methyl ester 95%
3-(2-Pyridinyl)-2-propenoic acid methyl ester 95%
3-(2-Pyridinyl)-2-propenoic acid methyl ester 95%
APOSOR955550-1G
:
Apollo Scientific
:
3-(2-Pyridinyl)-2-propenoic acid methyl ester 95%
APOSOR955550-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
163.173 g/mol
:
Ambient
:
50610-14-5
:
C₉H₉NO₂
:
MFCD18803084