5-(4-Chlorophenyl)-3H-1,3,4-oxadiazol-2-one 97%
Catalog # APOSOR954542-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.591 g/mol
Storage Temperature:Ambient
CAS Number:1711-61-1
Formula:C₈H₅ClN₂O₂
MDL Number:MFCD11046715