Order Entry
Italy
ContactUsLinkComponent
4,4,6-Trimethyl-1,3-dihydroquinolin-2-one 98%
Irritant
4,4,6-Trimethyl-1,3-dihydroquinolin-2-one 98%
Catalog # APOSOR954461-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4,4,6-Trimethyl-1,3-dihydroquinolin-2-one 98%
Catalog # APOSOR954461-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :189.254 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:181122-00-9
Formula:C₁₂H₁₅NO
MDL Number:MFCD12913845