Order Entry
Italy
ContactUsLinkComponent
Ethyl thiomorpholine-2-carboxylate 95%
Ethyl thiomorpholine-2-carboxylate 95%
Catalog # APOSOR952686-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl thiomorpholine-2-carboxylate 95%
Catalog # APOSOR952686-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :175.249 g/mol
Storage Temperature:Ambient
CAS Number:152009-44-4
Formula:C₇H₁₃NO₂S
MDL Number:MFCD20134212