6-Azaspiro[2.5]octan-4-ol hydrochloride
Catalog # APOSOR952020-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:163.645 g/mol
Storage Temperature:Ambient
CAS Number:955028-68-9
Formula:C₇H₁₃NO·ClH
MDL Number:MFCD21607169