Order Entry
Italy
ContactUsLinkComponent
(R)-2-Acetamido-2-phenylacetic acid 95%
(R)-2-Acetamido-2-phenylacetic acid 95%
Catalog # APOSOR949797-1G
Supplier:  Apollo Scientific
CAS Number:  
(R)-2-Acetamido-2-phenylacetic acid 95%
Catalog # APOSOR949797-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.199 g/mol
Storage Temperature:Ambient
CAS Number:14257-84-2
Formula:C₁₀H₁₁NO₃
MDL Number:MFCD12795908