Order Entry
Italy
ContactUsLinkComponent
3-(Bromomethyl)-1,1-dimethylcyclobutane
3-(Bromomethyl)-1,1-dimethylcyclobutane
Catalog # APOSOR948832-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Bromomethyl)-1,1-dimethylcyclobutane
Catalog # APOSOR948832-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.082 g/mol
Storage Temperature:Ambient
CAS Number:76207-22-2
Formula:C₇H₁₃Br
MDL Number:MFCD17170399