Order Entry
Italy
ContactUsLinkComponent
2,6-Dichloro-N-methylpyridin-4-amine 95%
2,6-Dichloro-N-methylpyridin-4-amine 95%
Catalog # APOSOR948201-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dichloro-N-methylpyridin-4-amine 95%
Catalog # APOSOR948201-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.031 g/mol
Storage Temperature:Ambient
CAS Number:175461-33-3
Formula:C₆H₆Cl₂N₂
MDL Number:MFCD12024871