Order Entry
Italy
ContactUsLinkComponent
Methyl 4-(methylamino)butanoate hydrochloride 95%
Methyl 4-(methylamino)butanoate hydrochloride 95%
Catalog # APOSOR947245-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 4-(methylamino)butanoate hydrochloride 95%
Catalog # APOSOR947245-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.634 g/mol
Storage Temperature:Ambient
CAS Number:89584-24-7
Formula:C₆H₁₃NO₂·ClH
MDL Number:MFCD11505402