Order Entry
Italy
ContactUsLinkComponent
2,2'-Hydrazobis[2-methylpropanenitrile] 95%
2,2'-Hydrazobis[2-methylpropanenitrile] 95%
Catalog # APOSOR947219-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2'-Hydrazobis[2-methylpropanenitrile] 95%
Catalog # APOSOR947219-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :166.224 g/mol
Storage Temperature:Ambient
CAS Number:6869-07-4
Formula:C₈H₁₄N₄
MDL Number:MFCD00045612