Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2-(1-methylpiperidin-4-ylidene)acetate
Methyl 2-(1-methylpiperidin-4-ylidene)acetate 95%
Methyl 2-(1-methylpiperidin-4-ylidene)acetate 95%
Methyl 2-(1-methylpiperidin-4-ylidene)acetate 95%
APOSOR942966-1G
:
Apollo Scientific
:
Methyl 2-(1-methylpiperidin-4-ylidene)acetate 95%
APOSOR942966-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
169.221 g/mol
:
Ambient
:
154594-25-9
:
C₉H₁₅NO₂
:
MFCD12756506