Order Entry
Italy
ContactUsLinkComponent
Ethyl 5-(cyanomethyl)furan-2-carboxylate 95%
Ethyl 5-(cyanomethyl)furan-2-carboxylate 95%
Catalog # APOSOR942839-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 5-(cyanomethyl)furan-2-carboxylate 95%
Catalog # APOSOR942839-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :179.173 g/mol
Storage Temperature:Ambient
CAS Number:51129-66-9
Formula:C₉H₉NO₃
MDL Number:MFCD12403566