
4,5,6,7-Tetrahydrofuro[3,2-c]pyridine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:150322-87-5
:
Formula:C₇H₉NO
MDL Number:MFCD09805956
:123.152 g/mol
Storage Temperature:Refrigerator
UN:3259
ADR:8,III
:GHS05
Frequently Bought Together