Order Entry
Italy
ContactUsLinkComponent
2,4,6,7-Tetrahydro-pyrano[4,3-c]pyrazole-3-carboxylic acid 95%
2,4,6,7-Tetrahydro-pyrano[4,3-c]pyrazole-3-carboxylic acid 95%
Catalog # APOSOR942428-1G
Supplier:  Apollo Scientific
CAS Number:  
2,4,6,7-Tetrahydro-pyrano[4,3-c]pyrazole-3-carboxylic acid 95%
Catalog # APOSOR942428-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.15 g/mol
Storage Temperature:Ambient
CAS Number:933694-89-4
Formula:C₇H₈N₂O₃
MDL Number:MFCD15202266