Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methoxybenzo[b]thiophene-2-carbaldehyde
5-Methoxybenzo[b]thiophene-2-carbaldehyde 95%
5-Methoxybenzo[b]thiophene-2-carbaldehyde 95%
APOSOR942338-1G
:
Apollo Scientific
:
5-Methoxybenzo[b]thiophene-2-carbaldehyde 95%
APOSOR942338-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
192.234 g/mol
:
Ambient
:
622864-56-6
:
C₁₀H₈O₂S
:
MFCD11101036