Order Entry
Italy
Orders LinkContactUsLinkComponent
Boc-DL-Leucinol 97%

Boc-DL-Leucinol 97%

Supplier:   Apollo Scientific
Boc-DL-Leucinol 97%
Supplier:  Apollo Scientific
Certificates
Boc-DL-Leucinol 97%

2 Options Available Below

Product Details & Documents
  • CAS Number:142121-48-0
  • Formula:C₁₁H₂₃NO₃
  • MDL Number:MFCD01883028
  • Molecular Weight:217.305 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 218,00 €
    218,00 €
    1 * 1 g
       218,00 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 549,00 €
    549,00 €
    1 * 5 g
       549,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.