Methyl 2-{3-azaspiro[5.5]undecan-9-yl}acetate hydrochloride
Catalog # APOSOR935882-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:261.788 g/mol
Storage Temperature:Ambient
CAS Number:1017540-91-8
Formula:C₁₃H₂₃NO₂·ClH
MDL Number:MFCD28501529