6,7-Dihydroisoquinolin-8(5H)-one hydrochloride 95%
Catalog # APOSOR932821-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:183.635 g/mol
Storage Temperature:Ambient
CAS Number:135311-97-6
Formula:C₉H₉NO·ClH
MDL Number:MFCD18432614