(R)-1-(2-Chlorophenyl)ethanamine hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1167414-92-7
:
Formula:C₈H₁₀ClN·ClH
MDL Number:MFCD11101198
:192.086 g/mol
Storage Temperature:Ambient
Frequently Bought Together