Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(2-Nitrophenyl)propan-1-ol
3-(2-Nitrophenyl)propan-1-ol 95%
3-(2-Nitrophenyl)propan-1-ol 95%
3-(2-Nitrophenyl)propan-1-ol 95%
APOSOR931377-1G
:
Apollo Scientific
:
3-(2-Nitrophenyl)propan-1-ol 95%
APOSOR931377-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
181.189 g/mol
:
Ambient
:
20716-26-1
:
C₉H₁₁NO₃
:
MFCD09028697