Calcium lactate gluconate 97%
Catalog # APOSOR931348-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: rac-(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, 2-hydroxypropanoate, Calcium
:
:324.295 g/mol
Storage Temperature:Ambient
CAS Number:11116-97-5
Formula:Ca·C₆H₁₁O₇·C₃H₅O₃
MDL Number:MFCD01320836
Melting Pt:140 °C