Ethyl l-thiazolidine-4-carboxylate hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:86028-91-3
:
Formula:C₆H₁₁NO₂S·ClH
MDL Number:MFCD09031280
:197.683 g/mol
Storage Temperature:Ambient
Frequently Bought Together