Order Entry
Italy
ContactUsLinkComponent
5,7-Dimethylpyrazolo[1,5-a]pyrimidine 95%
5,7-Dimethylpyrazolo[1,5-a]pyrimidine 95%
Catalog # APOSOR930461-1G
Supplier:  Apollo Scientific
CAS Number:  
5,7-Dimethylpyrazolo[1,5-a]pyrimidine 95%
Catalog # APOSOR930461-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :147.177 g/mol
Storage Temperature:Ambient
CAS Number:35149-38-3
Formula:C₈H₉N₃
MDL Number:MFCD01692984