Methyl 2-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:223788-08-7
:
Formula:C₈H₉NO₃
MDL Number:MFCD18803546
:167.162 g/mol
Storage Temperature:Ambient
Frequently Bought Together