C-(2-Methyl-imidazo[1,2-a]pyridin-3-yl)-methylamine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:34164-91-5
:
Formula:C₉H₁₁N₃
MDL Number:MFCD06739259
:161.204 g/mol
Storage Temperature:Ambient
Frequently Bought Together