Order Entry
Italy
ContactUsLinkComponent
7-Chloro-2H-1,4-benzoxazin-3(4h)-one 98%
7-Chloro-2H-1,4-benzoxazin-3(4h)-one 98%
Catalog # APOSOR926962-1G
Supplier:  Apollo Scientific
CAS Number:  
7-Chloro-2H-1,4-benzoxazin-3(4h)-one 98%
Catalog # APOSOR926962-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.592 g/mol
Storage Temperature:Ambient
CAS Number:27320-99-6
Formula:C₈H₆ClNO₂
MDL Number:MFCD03019820