Order Entry
Italy
ContactUsLinkComponent
Ethyl 3-(chloromethyl)-1H-pyrazole-4-carboxylate 95%
Ethyl 3-(chloromethyl)-1H-pyrazole-4-carboxylate 95%
Catalog # APOSOR925561-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 3-(chloromethyl)-1H-pyrazole-4-carboxylate 95%
Catalog # APOSOR925561-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.612 g/mol
Storage Temperature:Ambient
CAS Number:137487-60-6
Formula:C₇H₉ClN₂O₂
MDL Number:MFCD07357430