Order Entry
Italy
ContactUsLinkComponent
7-Methyl-7h-pyrrolo[2,3-d]pyrimidin-4-amine 95%
7-Methyl-7h-pyrrolo[2,3-d]pyrimidin-4-amine 95%
Catalog # APOSOR923975-1G
Supplier:  Apollo Scientific
CAS Number:  
7-Methyl-7h-pyrrolo[2,3-d]pyrimidin-4-amine 95%
Catalog # APOSOR923975-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :148.165 g/mol
Storage Temperature:Ambient
CAS Number:7752-54-7
Formula:C₇H₈N₄
MDL Number:MFCD12924275