Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dimethyl-1H-pyrrolo[2,3-b]pyridine
2,3-Dimethyl-1H-pyrrolo[2,3-b]pyridine 95%
2,3-Dimethyl-1H-pyrrolo[2,3-b]pyridine 95%
APOSOR923961-1G
:
Apollo Scientific
:
2,3-Dimethyl-1H-pyrrolo[2,3-b]pyridine 95%
APOSOR923961-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
146.189 g/mol
:
Ambient
:
10299-69-1
:
C₉H₁₀N₂
:
MFCD00513469