Order Entry
Italy
ContactUsLinkComponent
4-Chloro-7-isopropyl-7h-pyrrolo[2,3-d]pyrimidine 95%
4-Chloro-7-isopropyl-7h-pyrrolo[2,3-d]pyrimidine 95%
Catalog # APOSOR923112-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-7-isopropyl-7h-pyrrolo[2,3-d]pyrimidine 95%
Catalog # APOSOR923112-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.649 g/mol
Storage Temperature:Ambient
CAS Number:870706-47-1
Formula:C₉H₁₀ClN₃
MDL Number:MFCD21336906