5-Chloropyrazolo[1,5-a]pyrimidine-3-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1315364-91-0
:
Formula:C₇H₄ClN₃O₂
MDL Number:MFCD20482240
:197.579 g/mol
Storage Temperature:Ambient
Frequently Bought Together