
6-Chloro-1H-pyrrolo[3,2-c]pyridine-3-carbaldehyde 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1000341-64-9
:
Formula:C₈H₅ClN₂O
MDL Number:MFCD09749983
:180.591 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together