
3-(Chloromethyl)-1,5-dimethyl-1H-pyrazole 97%
Catalog # APOSOR9184-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:144.602 g/mol
Melting Pt:63…66 °C
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
CAS Number:180519-09-9
Formula:C₆H₉ClN₂
MDL Number:MFCD08060499