Order Entry
Italy
ContactUsLinkComponent
N-Ethyl-N-(2-pyrrolidinylmethyl)ethanamine 95%
N-Ethyl-N-(2-pyrrolidinylmethyl)ethanamine 95%
Catalog # APOSOR918256-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Ethyl-N-(2-pyrrolidinylmethyl)ethanamine 95%
Catalog # APOSOR918256-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :156.268 g/mol
Storage Temperature:Ambient
CAS Number:121053-95-0
Formula:C₉H₂₀N₂
MDL Number:MFCD06653367