3-Hydroxy-6,6-dimethyl-1-heptene-4-yne 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:78629-20-6
:
Formula:C₉H₁₄O
MDL Number:MFCD04974030
:138.207 g/mol
Storage Temperature:Ambient
UN:3082
ADR:9+EHS,III
Frequently Bought Together